Order Entry
Canada
Orders LinkContactUsLinkComponent
Pliers, Insertion/Extraction, Excelta

Pliers, Insertion/Extraction, Excelta

Pliers, insertion/extraction, 5 star quality, Carbon, 137.5 mm (5.5"), 4.5 mm (0.18"), 31.5 mm (1.260")
Catalog # 63042-444
Supplier:   Excelta
Supplier #:  505A-US
Pliers, Insertion/Extraction, Excelta
Catalog # 63042-444
Supplier:  Excelta
Pliers, Insertion/Extraction, Excelta
Pliers, Insertion/Extraction, Excelta

Pliers, insertion/extraction, 5 star quality, Carbon, 137.5 mm (5.5"), 4.5 mm (0.18"), 31.5 mm (1.260")

Price
$639.75
Log in for availability
Each
Product Details & Documents

These Insertion/Extraction pliers are designed to handle DIP/IC package.

  • Insertion, extraction pliers
  • Soft jaws eliminate damage to component
  • Ergonomic grips for comfort
  • Double leaf springs for ease of use

These soft tip insertion/extraction tools have soft acetal inserts eliminating scratching and nicking of delicate IC leads. The 'dash number' references the total amount of pins on package. It features a ergonomically molded antimicrobial grip (10¹⁰ ohms/sq surface resistivity) with dual leaf springs. The shanks are made of carbon tool steel.

Specifications
  • Description:
    Pliers, insertion/extraction, 5 star quality
  • Length:
    137.5 mm (5.5")
  • Material:
    Carbon
  • Jaw width:
    31.5 mm (1.260")
  • Jaw length:
    4.5 mm (0.18")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.