Order Entry
Canada
Orders LinkContactUsLinkComponent
Scissors, Trimmer, Pattern Cutting, Excelta Corp®

Scissors, Trimmer, Pattern Cutting, Excelta Corp®

Scissors, trimmer, 2 star quality, 6" (150 mm), Stainless steel AISI 440, 69.9 mm (2¾"), Straight
Catalog # 63041-974
Supplier:   Excelta
Supplier #:  339
Scissors, Trimmer, Pattern Cutting, Excelta Corp®
Catalog # 63041-974
Supplier:  Excelta
Scissors, Trimmer, Pattern Cutting, Excelta Corp®
Scissors, Trimmer, Pattern Cutting, Excelta Corp®

Scissors, trimmer, 2 star quality, 6" (150 mm), Stainless steel AISI 440, 69.9 mm (2¾"), Straight

Price
54,40 $
Log in for availability
Each
Product Details & Documents

These two star trimmer scissors are used for pattern cutting and general purpose use.

  • Ideal scissors for pattern cutting
  • Three different sizes to choose from

They are chrome-plated and come in various lengths.

Specifications
  • Description:
    Scissors, trimmer, 2 star quality
  • Length:
    6" (150 mm)
  • Material:
    Stainless steel AISI 440
  • Curvature:
    Straight
  • Blade length:
    69.9 mm (2¾")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.