Order Entry
Canada
Orders LinkContactUsLinkComponent
PYREX® Drying Trays, Corning

PYREX® Drying Trays, Corning

1750 ml, 27.2 x 18 x 5 cm (1023/32 x 7 x 2")
Catalog # 62670-010
Supplier:   Corning
Supplier #:  3175-7
PYREX® Drying Trays, Corning
Catalog # 62670-010
Supplier:  Corning
PYREX® Drying Trays, Corning
PYREX® Drying Trays, Corning

1750 ml, 27.2 x 18 x 5 cm (1023/32 x 7 x 2")

Price
$139.12
New Customer Promotional Price
Log in for availability
Case of 4
Product Details & Documents

These dishes made of PYREX® borosilicate glass will withstand hot air or steam sterilization and are useful for drying, staining or developing trays.

  • Excellent chemical resistance
  • Good temperature resistance
  • Easy to clean

They can be used with gel electrophoresis or paper chromatography.

Caution: For oven use only. Do not use over direct heat.

Specifications
  • Volume:
    1750 ml
  • L×W×H:
    27.2 x 18 x 5 cm (1023/32 x 7 x 2")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.