Trypsinizing Flasks, with Cap, Wheaton
700 ml, 51-400
Catalog # 62401-877
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: 355807
Jump to:
Product Details & Documents
Wheaton-33® low extractable borosilicate glass composition with narrow lip
Deep uniform baffles provide excellent turbulence. Internal dam reduces spill out of untrypsinized particles. Black screw cap with white (styrene-butadiene) rubber liner.
Specifications
- Cap Size:51-400
- Capacity:700 ml