Trypsinizing Flasks, with Cap, Wheaton
150 ml, 38-430
Catalog # 62401-640
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: 355444
Jump to:
Product Details & Documents
Wheaton-33® low extractable borosilicate glass composition with narrow lip
Deep uniform baffles provide excellent turbulence. Internal dam reduces spill out of untrypsinized particles. Black screw cap with white (styrene-butadiene) rubber liner.
Specifications
- Cap Size:38-430
- Capacity:150 ml