Orders LinkContactUsLinkComponent
Trypsinizing Flasks, with Cap, Wheaton
Trypsinizing Flasks, with Cap, Wheaton
1 L, 51-400
Catalog # 62401-632
Supplier #:  355397
Trypsinizing Flasks, with Cap, Wheaton
Catalog # 62401-632
Trypsinizing Flasks, with Cap, Wheaton
Trypsinizing Flasks, with Cap, Wheaton
1 L, 51-400
Price
$167.44
New Customer Promotional Price
Log in for availability
Case of 1
Product Details & Documents
Wheaton-33® low extractable borosilicate glass composition with narrow lip
Deep uniform baffles provide excellent turbulence. Internal dam reduces spill out of untrypsinized particles. Black screw cap with white (styrene-butadiene) rubber liner.
Specifications
  • Cap Size:
    51-400
  • Capacity:
    1 L