Erlenmeyer Shake Flasks with Three Baffles, Wheaton
125 ml
Catalog # 62401-575
Supplier: DWK Life Sciences (KIMBLE)
Supplier #: 353257
Erlenmeyer Shake Flasks with Three Baffles, Wheaton
Catalog # 62401-575
Supplier: DWK Life Sciences (KIMBLE)
Jump to:
Product Details & Documents
Designed for use on either rotary or reciprocating shakers.
- Autoclavable
- Specially designed to maximize aeration for improved growth
- Pinched neck design to reduce splashing on flasks with four baffles
Feature a marking spot on the side of the flask and a break-resistant lip, which accommodates a cotton plug or stopper. Made of wheaton 33® low-extractable borosilicate glass, which meets ASTM Type I Class A and USP Type I standards.
Documents
Specifications
- Capacity:125 ml