Five-Channel Timer, Sper Scientific
5 channel timer, 2⅞×2½×½", 2 oz.
Catalog # 62364-185
Supplier: Sper Scientific
: 810015
Jump to:
Product Details & Documents
This digital count down/count up timer has four independent channels, each of which counts up or down for any interval from 1 second to 100 hours, plus clock button.
- Extra large display indicates channels in use
- Each one has a distinctive 1 minute alarm
Automatically begins to count up to indicate time elapsed after count down reaches zero. Memory recalls last timer setting or reuse. Also has a clock mode. Has a stand, pocket clip, and magnet.
Certifications: N.I.S.T. traceable certificate of compliance for the certified option.
Specifications
- Description:5 channel timer
- W×H×D:2⅞×2½×½"
- Weight:2 oz.