Order Entry
Canada
ContactUsLinkComponent
 
VWR® Mini-Alarm Timer/Stopwatch
  62344-585
undefined
VWR® Mini-Alarm Timer/Stopwatch
  62344-585

 

  • Environmentally Preferable:
  • Cat. no.:
    62344-585
  • H×W×D:
    6.3×5.6×1.3 cm (2¹/₂×2¹/₄×¹/₂")

 

 

Use individually numbered keys to quickly set countdown time to any value up to 99 minutes, 99 seconds.

  • Traceable®
  • LCD digits are readable from 2.4 m (8') away
  • Rugged, chemical-resistant ABS plastic case

Stopwatch mode with time-out feature counts up to 59 minutes, 59 seconds, then automatically rolls over to zero to continue timing.

Unit incorporates a flip-open easel for benchtop use, a spring fastener for clipping to lab coat, and a magnetic back for use with incubators, refrigerators, ovens, and more.

High decibel countdown alarm turns off manually or automatically after one minute.

 : A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
 : Supplied with a two-year, 1.5 V silver-oxide battery.
Frequently Bought Together