Order Entry
Canada
Orders LinkContactUsLinkComponent
Data-Trend Multipurpose Temperature Chart Recorders, Dickson®

Data-Trend Multipurpose Temperature Chart Recorders, Dickson®

7-Day Chart, 0...250 °C/°F
Catalog # 61280-554
Supplier:   Dickson
Supplier #:  C659
Data-Trend Multipurpose Temperature Chart Recorders, Dickson®
Catalog # 61280-554
Supplier:  Dickson
Data-Trend Multipurpose Temperature Chart Recorders, Dickson®
Data-Trend Multipurpose Temperature Chart Recorders, Dickson®

7-Day Chart, 0...250 °C/°F

Price
$192.29
Log in for availability
Each Pack of 60
Product Details & Documents

Adaptable to a wide range of applications, these recorders feature a 15.2cm (6") readable chart, AC power with battery backup, user-selectable recording times and temperature ranges, and a remote probe

Increased shielding eliminates electronic noise interference and allows smoother pen movement.

Ordering information: Recorders include red pen, AC adapter, 9V battery, and 1.2m (4') bead wire probe. Charts must be purchased separately.

Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.

Specifications
  • Temperature range:
    0...250 °C/°F
  • Description:
    7-Day Chart
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Red Pens
    Red Pens
    61280-132
    Log in for availability
    Pack of 6 - $244.14

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.