Orders LinkContactUsLinkComponent
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
8 L (2 gal.), 20 x 20 x 20 cm (8 x 8 x 8")
Catalog # 60463-166
Supplier #:  14100-0002
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Catalog # 60463-166
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
8 L (2 gal.), 20 x 20 x 20 cm (8 x 8 x 8")
Price
309,60 $
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
Tanks have stepped rim for strength and overlapping skirted cover
Molded in one piece with good chemical resistance
 : Tanks can be modified with factory-installed spigots; contact your Avantor sales representative for more information.
Specifications
  • Volume:
    8 L (2 gal.)
  • L×W×D:
    20 x 20 x 20 cm (8 x 8 x 8")