Order Entry
Canada
Orders LinkContactUsLinkComponent
Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific

Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific

20 L (5 gal.), 25 x 25 x 36 cm (10 x 10 x 14")
Catalog # 60463-107
Supplier #:  14200-0005
Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific
Catalog # 60463-107
Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific
Nalgene® Rectangular Tanks, Polypropylene, Thermo Scientific

20 L (5 gal.), 25 x 25 x 36 cm (10 x 10 x 14")

Price
$584.00
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

Tanks have stepped rim for strength and overlapping skirted cover

Polypropylene tank is molded in one piece and is autoclavable

Certifications: Plastic meets USP VI and FDA 21 CFR 177.1520 standards.

Ordering information: Tanks can be modified with factory-installed spigots; contact your Avantor sales representative for more information.

Documents

Specifications
  • Volume:
    20 L (5 gal.)
  • L×W×D:
    25 x 25 x 36 cm (10 x 10 x 14")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.