Orders LinkContactUsLinkComponent
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
120 L (32 gal.), 61 x 45 x 45 cm (24 x 18 x 18")
Catalog # 60463-084
Supplier #:  14100-0045
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Catalog # 60463-084
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
120 L (32 gal.), 61 x 45 x 45 cm (24 x 18 x 18")
Price
$864.00
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
Tanks have stepped rim for strength and overlapping skirted cover
Molded in one piece with good chemical resistance
Ordering information: Tanks can be modified with factory-installed spigots; contact your Avantor sales representative for more information.
Specifications
  • Volume:
    120 L (32 gal.)
  • L×W×D:
    61 x 45 x 45 cm (24 x 18 x 18")
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.