Order Entry
Canada
Orders LinkContactUsLinkComponent
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific

Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific

38 L (10 gal.), 46 x 30 x 30 cm (18 x 12 x 12")
Catalog # 60463-040
Supplier #:  14100-0015
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Catalog # 60463-040
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific

38 L (10 gal.), 46 x 30 x 30 cm (18 x 12 x 12")

Price
510,40 $
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents

Tanks have stepped rim for strength and overlapping skirted cover

Molded in one piece with good chemical resistance

Ordering information: Tanks can be modified with factory-installed spigots; contact your Avantor sales representative for more information.

Specifications
  • Volume:
    38 L (10 gal.)
  • L×W×D:
    46 x 30 x 30 cm (18 x 12 x 12")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.