Orders LinkContactUsLinkComponent
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
30 L (8 gal.), 30 x 30 x 30 cm (12 x 12 x 12")
Catalog # 60463-029
Supplier #:  14100-0010
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Catalog # 60463-029
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
Nalgene® Rectangular Tanks, Low-Density Polyethylene, Thermo Scientific
30 L (8 gal.), 30 x 30 x 30 cm (12 x 12 x 12")
Price
$390.40
New Customer Promotional Price
Log in for availability
Each
Product Details & Documents
Tanks have stepped rim for strength and overlapping skirted cover
Molded in one piece with good chemical resistance
Ordering information: Tanks can be modified with factory-installed spigots; contact your Avantor sales representative for more information.
Specifications
  • Volume:
    30 L (8 gal.)
  • L×W×D:
    30 x 30 x 30 cm (12 x 12 x 12")