VWR® Talon® Extra-Large Lab-Frames
Lab-Frame with Aluminum Rods, Aluminum
Catalog # 60078-006
Supplier: VWR International
Jump to:
Product Details & Documents
Extra-large lab frames offer the greatest versatility for your most demanding setups.
- Extra large design
- For horizontal or vertical use
- Versatile and adaptable
- Mountable on either side
Measure 1220Lx1830Hmm (48x72"). May be used either horizontally or vertically. Include six 305mm (12") rods and feet for securing frame to wall. Apparatus mountable on either side of frame.
Ordering information: Each Lab Frame includes six 51 mm (2") rods, six 305 mm (12") rods, two 610 mm (24") rods, seven 1220 mm (48") rods, five 1829 mm (72") rods, four rod end connectors, forty-five S-connectors, and eight Lab Frame feet.
Documents
Lab-Frame 4
Manual
VWR Laboratory Equipments
Product Literature
Specifications
- Description:Lab-Frame with Aluminum Rods
- Material:Aluminum