Synthware Sure-Grip Separatory Funnels
With glass stopper, 500 ml, 4 mm, 27
60004-246
: F687500A
Globe-shaped separatory funnel features a long stopper neck and a straight section between the funnel body and stopcock that provides a secure grip while shaking
- Sure-grip design
- Heavy wall borosilicate construction
- Ground glass joint
The straight section also aids in clamping. Available with PTFE stopcocks. Funnels are supplied complete with one PTFE or hollow glass stopper.
- Capacity:500 ml
- Description:With glass stopper
- Stopper no.:27
- Stopcock size:4 mm



