Synthware Flask, Filtering, Microscale, Kemtech America
50 ml
Catalog # 60004-226
Supplier: Kemtech America
Supplier #: F150050
Synthware Flask, Filtering, Microscale, Kemtech America
Catalog # 60004-226
Supplier: Kemtech America
Jump to:
Product Details & Documents
Filtering flask for microscale applications.
- Microscale
- Heavy wall Borosilicate construction
- Beaded top rim
- Barbed side arm
Top is tooled for a #2 neoprene filter adapter or #3 rubber stopper.
Specifications
- Capacity:50 ml