Order Entry
Canada
Orders LinkContactUsLinkComponent
Synthware Flasks, Erlenmeyer, Threaded, Microscale, Kemtech America

Synthware Flasks, Erlenmeyer, Threaded, Microscale, Kemtech America

25 ml, 14/10
Catalog # 60004-222
Supplier:   Kemtech America
Supplier #:  F501025
Synthware Flasks, Erlenmeyer, Threaded, Microscale, Kemtech America
Catalog # 60004-222
Supplier:  Kemtech America
Synthware Flasks, Erlenmeyer, Threaded, Microscale, Kemtech America
Synthware Flasks, Erlenmeyer, Threaded, Microscale, Kemtech America

25 ml, 14/10

Price
$39.26
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Erlenmeyer threaded flask for microscale applications.

  • Heavy wall borosilicate construction
  • Threaded upper joint
  • Open cap included

Supplied with a nylon compression cap, Viton® O-ring, and PTFE-lined silicone septum.

Specifications
  • [ST] joint no.:
    14/10
  • Capacity:
    25 ml
Accessories

Product Information

  • Catalog #
    I.D.×W
    O-Ring Size
    Availability
    Price
  • Viton® O-Ring, 14/10 threaded joints
    Viton® O-Ring, 14/10 threaded joints
    60004-998
    014
    Log in for availability
    Each - $5.61

Product Information

  • Catalog #
    Material
    Cap Size
    Availability
    Price
  • Nylon cap
    Nylon cap
    60005-054
    Nylon
    20 mm
    Log in for availability
    Each - $4.65

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.