Synthware Microscale Round-Bottom Flasks, Threaded, Kemtech America
25 ml, 14/10
Catalog # 60004-186
Supplier: Kemtech America
Supplier #: F301025
Synthware Microscale Round-Bottom Flasks, Threaded, Kemtech America
Catalog # 60004-186
Supplier: Kemtech America
Jump to:
Product Details & Documents
Supplied with one compression cap, one Viton® O-ring, and one fluoropolymer resin-faced silicone rubber septum.
- Threaded neck
- Ground glass joint
- Heavy wall borosilicate construction
- Various capacities and joint sizes available
- Cap included
Threaded ground glass joint allows for the use of a threaded cap or a ground glass stopper.
Ordering information: For replacement caps, contact your Avantor representative.
Specifications
- [ST] joint no.:14/10
- Capacity:25 ml