Orders LinkContactUsLinkComponent
Synthware Round-Bottom Flasks with Four Necks
Synthware Round-Bottom Flasks with Four Necks
1000 ml, 29/42, 24/40
Catalog # 60002-984
Supplier:   Kemtech America
Supplier #:  F21841L
Synthware Round-Bottom Flasks with Four Necks
Catalog # 60002-984
Supplier:  Kemtech America
Synthware Round-Bottom Flasks with Four Necks
Synthware Round-Bottom Flasks with Four Necks
1000 ml, 29/42, 24/40
Price
$356.45
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
With identical vertical side joints.
  • 3 Straight side necks
  • Ground glass joints
  • Heavy wall borosilicate construction
  • Various capacities and joint sizes available
Hand-blown from tubing to ensure heavy, uniform wall.
Specifications
  • Capacity:
    1000 ml
  • Center neck [ST] joint:
    29/42
  • Side neck ST joints:
    24/40