Orders LinkContactUsLinkComponent
Synthware Round-Bottom Flasks with Three Necks, Angled
Synthware Round-Bottom Flasks with Three Necks, Angled
500 ml, 24/40, 19/22
Catalog # 60002-734
Supplier:   Kemtech America
Supplier #:  F430500
Synthware Round-Bottom Flasks with Three Necks, Angled
Catalog # 60002-734
Supplier:  Kemtech America
Synthware Round-Bottom Flasks with Three Necks, Angled
Synthware Round-Bottom Flasks with Three Necks, Angled
500 ml, 24/40, 19/22
Price
$191.94
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Round bottom with [ST] joints.
  • 3 Angled necks
  • Ground glass joints
  • Heavy wall Borosilicate construction
  • Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
Specifications
  • Capacity:
    500 ml
  • Center neck [ST] joint:
    24/40
  • Side neck ST joints:
    19/22