Orders LinkContactUsLinkComponent
Synthware Round-Bottom Flasks with Three Necks, Angled
Synthware Round-Bottom Flasks with Three Necks, Angled
500 ml, 19/22, 19/22
  60002-732
Supplier #:  F439500
Synthware Round-Bottom Flasks with Three Necks, Angled
  60002-732
Synthware Round-Bottom Flasks with Three Necks, Angled
Synthware Round-Bottom Flasks with Three Necks, Angled
500 ml, 19/22, 19/22
$177.63
Each
Minimum order quantity is 1
Round bottom with [ST] joints.
  • 3 Angled necks
  • Ground glass joints
  • Heavy wall Borosilicate construction
  • Various capacities and joint sizes available
Heavy wall design. Hand-blown from tubing to ensure uniform wall thickness.
  • Capacity:
    500 ml
  • Center neck [ST] joint:
    19/22
  • Side neck ST joints:
    19/22