Orders LinkContactUsLinkComponent
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
100 ml, 14/20, 14/20
Catalog # 60002-606
Supplier:   Kemtech America
Supplier #:  F411100
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
Catalog # 60002-606
Supplier:  Kemtech America
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
Synthware Round-Bottom Flasks with Two Necks, Heavy Wall
100 ml, 14/20, 14/20
Price
$114.98
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Round-bottom flasks with one straight [ST] joint and one angled [ST] joint
  • Angled side neck
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Various sizes available
Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Specifications
  • Capacity:
    100 ml
  • Center neck [ST] joint:
    14/20
  • Side neck ST joints:
    14/20