Orders LinkContactUsLinkComponent
Synthware Extraction Thimbles, Glass
Synthware Extraction Thimbles, Glass
Medium extractor size, 35×90 mm, Extra coarse
Catalog # 60002-220
Supplier:   Kemtech America
Supplier #:  E113590X
Synthware Extraction Thimbles, Glass
Catalog # 60002-220
Supplier:  Kemtech America
Synthware Extraction Thimbles, Glass
Synthware Extraction Thimbles, Glass
Medium extractor size, 35×90 mm, Extra coarse
Price
$97.64
New Customer Promotional Price
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
All-glass extraction thimbles permit viewing extraction process and subsequent weighing.
  • Heavy wall borosilicate construction
  • Coarse or extra coarse porosity available
Ordering information: Small thimble is used with condenser 60002-198 and extractor 60002-204; Medium thimble is used with condenser 60002-200 and extractors 60002-206 or 60002-208; Large thimble is used with condenser 60002-202 and extractors 60002-210 or 60002-212.
Specifications
  • Porosity:
    Extra coarse
  • O.D.×H:
    35×90 mm
  • Fits:
    Medium extractor size
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.