Synthware Extraction Thimbles, Glass
Medium extractor size, 35×90 mm, Coarse
Catalog # 60002-218
Supplier: Kemtech America
Supplier #: E113590C
Jump to:
Product Details & Documents
All-glass extraction thimbles permit viewing extraction process and subsequent weighing.
- Heavy wall borosilicate construction
- Coarse or extra coarse porosity available
Ordering information: Small thimble is used with condenser 60002-198 and extractor 60002-204; Medium thimble is used with condenser 60002-200 and extractors 60002-206 or 60002-208; Large thimble is used with condenser 60002-202 and extractors 60002-210 or 60002-212.
Specifications
- Porosity:Coarse
- O.D.×H:35×90 mm
- Fits:Medium extractor size
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.