Synthware Extraction Thimbles, Glass
Small extractor size, 25×85 mm, Extra coarse
Catalog # 60002-216
Supplier: Kemtech America
Supplier #: E112585X
Jump to:
Product Details & Documents
All-glass extraction thimbles permit viewing extraction process and subsequent weighing.
- Heavy wall borosilicate construction
- Coarse or extra coarse porosity available
Ordering information: Small thimble is used with condenser 60002-198 and extractor 60002-204; Medium thimble is used with condenser 60002-200 and extractors 60002-206 or 60002-208; Large thimble is used with condenser 60002-202 and extractors 60002-210 or 60002-212.
Specifications
- Porosity:Extra coarse
- O.D.×H:25×85 mm
- Fits:Small extractor size