Synthware Anti-Splash Adapters
250 ml, [ST] 29/42, 29/42
Catalog # 60000-430
Supplier: Kemtech America
Supplier #: A288250
Jump to:
Product Details & Documents
Anti-splash adapters are designed to prevent possible contamination from a rotary evaporator.
- Top outer [ST] joint
- Bottom inner [ST] joint
The upper joint is an outer [ST] joint, while the lower joint is an inner [ST] joint.
Specifications
- Capacity:250 ml
- Bottom [ST] joint size:29/42
- Top [ST] joint size:[ST] 29/42