Synthware Anti-Splash Adapters
100 ml, [ST] 29/42, 24/40
Catalog # 60000-418
Supplier: Kemtech America
Supplier #: A282924
Jump to:
Product Details & Documents
Anti-splash adapters are designed to prevent possible contamination from a rotary evaporator.
- Top outer [ST] joint
- Bottom inner [ST] joint
The upper joint is an outer [ST] joint, while the lower joint is an inner [ST] joint.
Specifications
- Capacity:100 ml
- Bottom [ST] joint size:24/40
- Top [ST] joint size:[ST] 29/42