Synthware Adapters with Ball Socket
Adapter, with ball socket, [ST] 35/20, 14/20
Catalog # 60000-090
Supplier: Kemtech America
Supplier #: A663514
Jump to:
Product Details & Documents
Ball socket adapter with lower standard taper inner joint.
- Heavy wall construction
- 33 expansion Borosilicate glass
- Standard ground glass joints work with other brands
For use with any 35/20 ball socket joint.
Specifications
- Description:Adapter, with ball socket
- Bottom [ST] joint size:14/20
- Top [ST] joint size:[ST] 35/20