Order Entry
Canada
Orders LinkContactUsLinkComponent
Wire Workstation, Mobile, Eagle MHC

Wire Workstation, Mobile, Eagle MHC

Wire Workstation with Two Cantilever Wire Overshelves, 48", 24", Chrome
Catalog # 58202-900
Supplier:   Eagle Group
Supplier #:  MWS2448WT
Wire Workstation, Mobile, Eagle MHC
Catalog # 58202-900
Supplier:  Eagle Group
Wire Workstation, Mobile, Eagle MHC
Wire Workstation, Mobile, Eagle MHC

Wire Workstation with Two Cantilever Wire Overshelves, 48", 24", Chrome

Price
$2,487.95
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents

Mobile wire workstation units offer 62% laminar flow and a 24" wide wire shelving workstation top

Four 33" legs and C-frame for additional stability. Features four 5" poly stem casters, two with brake. Weight capacity of up to 800lbs. evenly distributed. All units have standard working height of 33".

Documents

Specifications
  • Width:
    24"
  • Description:
    Wire Workstation with Two Cantilever Wire Overshelves
  • Length:
    48"
  • Finish:
    Chrome
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Stainless steel keyboard drawer
    Stainless steel keyboard drawer
    58202-976
    Log in for availability
    Each - $1,304.58

Product Information

  • Catalog #
    Availability
    Price
  • Mouse shelf
    Mouse shelf
    58202-978
    Log in for availability
    Each - $39.54

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.