Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Wire Workstation, Mobile, Eagle MHC

Wire Workstation with Two Cantilever Wire Overshelves, 36", 24", Chrome
Catalog # 58202-894
Supplier:   Eagle Group
 :  MWS2436WT
Wire Workstation, Mobile, Eagle MHC
Catalog # 58202-894
Supplier:  Eagle Group
Wire Workstation, Mobile, Eagle MHC
Wire Workstation, Mobile, Eagle MHC

Wire Workstation with Two Cantilever Wire Overshelves, 36", 24", Chrome

Price
$2,310.39
Log in for availability
Each
 
Product Details & Documents

Mobile wire workstation units offer 62% laminar flow and a 24" wide wire shelving workstation top

Four 33" legs and C-frame for additional stability. Features four 5" poly stem casters, two with brake. Weight capacity of up to 800lbs. evenly distributed. All units have standard working height of 33".

Specifications
  • Width:
    24"
  • Description:
    Wire Workstation with Two Cantilever Wire Overshelves
  • Length:
    36"
  • Finish:
    Chrome
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Stainless steel keyboard drawer
    Stainless steel keyboard drawer
    58202-976
    Log in for availability
    Each - $1,304.58

Product Information

  • Catalog #
    Availability
    Price
  • Mouse shelf
    Mouse shelf
    58202-978
    Log in for availability
    Each - $39.54