Drum Tray, Eagle Manufacturing
Drum Tray Dolly
Catalog # 56618-538
Supplier: Eagle Manufacturing
Supplier #: 1618
Jump to:
Product Details & Documents
Tray with reservoir keeps container away from spills
Ideal for use under drums during dispensing/filling operations. Holds up to 34L (9gal.) of spills when used with a 208.2L (55gal.) drum. Constructed of high-density polyethylene. Use drum tray dolly to easily transport drums between work areas without compromising spill protection.
Specifications
- Description:Drum Tray Dolly