Order Entry
Canada
Orders LinkContactUsLinkComponent
Cryoscope Standards, Advanced Instruments

Cryoscope Standards, Advanced Instruments

Cryoscope Standard 621 m°H, 10 x 10 mL ampules
Catalog # 55005-003
Supplier #:  3LA033
Cryoscope Standards, Advanced Instruments
Catalog # 55005-003
CAS Number:  
Cryoscope Standards, Advanced Instruments

Cryoscope Standard 621 m°H, 10 x 10 mL ampules

CAS Number:  
Price
279,24 $
Log in for availability
Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Cryoscope standards are used to calibrate and/or check the performance of a cryoscope.

Standards should only be used in accordance with the cryoscope being calibrated. Refer to the specific instrument's user guide for calibration instructions. These standards are prepared from salts. 2-Hydroxybiphenyl Sodium Salt Tetrahydrate, 0.01%, is added as a preservative.

Documents

Specifications

Specifications

  • Description:
    Cryoscope Standard 621 m°H, 10 x 10 mL ampules

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.