Generators for Polytron® PT Models, Kinematica
PT-DA 07 / 2SYN-E082, Sterile Disposable, 25/Pack, 0.30 - 10 ml, 7 mm, 7×82 mm, PT 1200 E PT 1300 D PT 2500 E
47751-632
- Ø:7 mm
- Description:PT-DA 07 / 2SYN-E082, Sterile Disposable, 25/Pack
- Diameter ×L:7×82 mm
- For use with:PT 1200 E PT 1300 D PT 2500 E
- Sample volume:0.30 - 10 ml



