Order Entry
Canada
Orders LinkContactUsLinkComponent
Technik™ Chain Nose Pliers

Technik™ Chain Nose Pliers

Chain Nose Pliers, 11.4 cm (4½"), 2.4 cm (¹⁵⁄₁₆"), Serrated
Catalog # 47743-084
Supplier:   Aven Tools
Supplier #:  10307
Technik™ Chain Nose Pliers
Catalog # 47743-084
Supplier:  Aven Tools
Technik™ Chain Nose Pliers
Technik™ Chain Nose Pliers

Chain Nose Pliers, 11.4 cm (4½"), 2.4 cm (¹⁵⁄₁₆"), Serrated

Price
$98.16
Log in for availability
Each
Product Details & Documents

Pliers feature dual leaf springs and box joint construction for comfortable action.

  • Made of mirror polished stainless steel
  • Induction hardened cutting edges
  • ESD-safe grips
Specifications
  • Description:
    Chain Nose Pliers
  • Length:
    11.4 cm (4½")
  • Jaw type:
    Serrated
  • Jaw length:
    2.4 cm (¹⁵⁄₁₆")
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.