Order Entry
Canada
Orders LinkContactUsLinkComponent
Germicidal Bulbs

Germicidal Bulbs

Bulb, Linear Single Ended, Voltarc, T5, 16 W, 253.7 nm, S400/4-pin, 7000 h, 357 mm, 15.875 mm
Catalog # 47736-302
Germicidal Bulbs
Catalog # 47736-302
Supplier:  
Germicidal Bulbs
Germicidal Bulbs

Bulb, Linear Single Ended, Voltarc, T5, 16 W, 253.7 nm, S400/4-pin, 7000 h, 357 mm, 15.875 mm

Discontinued
Product Details & Documents

Various germicidal bulbs.

Specifications
  • Ø:
    15.875 mm
  • Description:
    Bulb, Linear Single Ended, Voltarc
  • Length:
    357 mm
  • Bulb life:
    7000 h
  • Base:
    S400/4-pin
  • Model:
    T5
  • Power:
    16 W
  • Wavelength:
    253.7 nm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.