Order Entry
Canada
Orders LinkContactUsLinkComponent
Germicidal Bulbs

Germicidal Bulbs

Bulb, Linear Double Ended, Sankyo Denki, Rapid Start (Circuitry), T10, 40 W, 253.7 nm, G13/medium 2-pin, 8000 h, 1198 mm, 32.5 mm
Catalog # 47736-198
Supplier #:  0000179
Germicidal Bulbs
Catalog # 47736-198
Supplier:  
Germicidal Bulbs
Germicidal Bulbs

Bulb, Linear Double Ended, Sankyo Denki, Rapid Start (Circuitry), T10, 40 W, 253.7 nm, G13/medium 2-pin, 8000 h, 1198 mm, 32.5 mm

Discontinued
Product Details & Documents

Various germicidal bulbs.

Specifications
  • Ø:
    32.5 mm
  • Description:
    Bulb, Linear Double Ended, Sankyo Denki, Rapid Start (Circuitry)
  • Length:
    1198 mm
  • Bulb life:
    8000 h
  • Base:
    G13/medium 2-pin
  • Model:
    T10
  • Power:
    40 W
  • Wavelength:
    253.7 nm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.