Order Entry
Canada
Orders LinkContactUsLinkComponent
VWR® Basic SPP Frocks

VWR® Basic SPP Frocks

White, L, 43½", 35½"
Catalog # 414004-383
Supplier:   VWR International
VWR® Basic SPP Frocks
Catalog # 414004-383
Supplier:  VWR International
VWR® Basic SPP Frocks
VWR® Basic SPP Frocks

White, L, 43½", 35½"

Price
$313.57
New Customer Promotional Price
Log in for availability
Case of 30 Pallet of 720
Product Details & Documents

Disposable frock made from spunbond polypropylene (SPP) fabric, offering lightweight and breathable protection for non-hazardous environments. Designed for comfort and contamination control, it features snap-front closure, non-linting material, and an economical single-use design.

  • Lightweight and breathable fabric reduces heat buildup during long shifts

    Non-linting material helps maintain clean environments

    Snap-front closure for easy donning and removal

    Elastic cuffs for flexibility

    Extended coverage compared to lab coats

    Disposable, single-use design for contamination control

    Economical solution for basic protection needs

The VWR® Basic Protection Frock is designed for environments where cleanliness and contamination control are essential, but exposure risks remain minimal. Made from spunbond polypropylene (SPP), this non-woven fabric offers a lightweight and breathable solution that ensures comfort during extended wear. Its non-linting properties help reduce particle contamination, making it suitable for clean areas, food processing, and general industrial applications.

The frock features a snap-front closure for quick and secure fastening, along with elastic cuffs to accommodate different glove types and ensure a secure fit. The disposable design supports hygiene compliance by minimizing cross-contamination and simplifying inventory management. While not intended for hazardous chemical or liquid protection, the frock provides an effective barrier against dust, dirt, and dry particulates, making it ideal for routine tasks in controlled environments. Combining cost-effectiveness, breathability, and basic contamination control, VWR® Basic Protection SPP Frocks deliver a practical solution for facilities seeking dependable single-use apparel for everyday operations.

Packaging: Frocks are packaged 30 per case.

Specifications
  • Clothing Size:
    L
  • Color:
    White
  • Length:
    43½"
  • Sleeve:
    35½"
  • Sleeve or leg length:
    Long
  • Gender:
    Unisex
  • Closure type:
    Press studs / snaps
  • Water resistance:
    21.7 G
  • Tensile strength:
    25.9 lbs./in. (warp), 23.1 lbs./in. (filling)
  • Weight:
    48.82 g/m²
  • Bacterial filtration efficiency:
    55%
  • Tearing strength:
    4.2 lbs./in. (lengthwise yarns), 4 lbs./in. (widthwise yarns)
  • Cat. No.:
    414004-383
  • Cleanroom class:
    10000/ISO 7
  • Air permeability:
    840.63 cu. ft./min/sq. ft.
  • Garment application:
    Cleanroom|Industrial|Laboratory
  • Pockets:
    Without pockets
  • Fabric:
    Spunbound polypropylene
  • Particle shedding (Helmke Drum):
    Level II
  • Water vapor transmission rate:
    1257 g/m²/24 h