VWR® Basic SMS Coveralls
Product Details & Documents
VWR® Basic Protection SMS Coveralls are manufactured from a latex-free, multi-layer spunbonded-meltblown-spunbonded (SMS) fabric that offers high tensile strength while remaining soft, comfortable, and breathable. These static dissipative garments act as a fluid and particulate barrier and provide light chemical splash resistance.
SMS (Spunbond-Meltblown-Spunbond) nonwoven fabric
- Multi-layer protection combines strength, filtration, and comfort
- Outer spunbond layers allow airflow while maintaining barrier integrity
- Middle meltblown layer provides enhanced protection against particulates and limited liquid splashes
- Zipper front closure, elastic wrists and ankles
- Tear-resistant and abrasion-resistant for extended wear
- Effective barrier against dust, dry particulates, and light liquid splashes
- Breathable design reduces heat stress during long shifts
- Meets industry standards for nonhazardous protective apparel
The SMS coverall is engineered for environments requiring enhanced protection without sacrificing comfort. Constructed from SMS fabric (Spunbond-Meltblown-Spunbond), this material combines three layers to deliver superior performance. The outer spunbond layers provide strength and durability, while the meltblown middle layer acts as an effective barrier against particulates and limited liquid splashes. This multi-layer design ensures a balance of breathability, protection, and flexibility, making SMS coveralls ideal for extended wear in controlled environments.
Key features include a zipper front closure for secure fit, and elastic wrists and ankles to minimize exposure gaps. SMS coveralls are non-linting, reducing contamination risks, and are widely used in pharmaceutical, cleanroom, food processing, and industrial settings where particulate control and splash resistance are critical.
Unlike basic polypropylene garments, SMS coveralls offer higher barrier protection while maintaining comfort, making them suitable for tasks involving powders, dry particulates, and light liquid contact. Their disposable nature supports hygiene compliance and simplifies inventory management, providing a practical and cost-effective solution for facilities prioritizing both safety and efficiency.
Packaging: Coveralls are packaged 25 per case.
Specifications
- Clothing Size:L
- Color:White
- Length:38¾" (body), 29½" (leg)
- Chest size:25"
- Sleeve:34"
- Sleeve or leg length:Long
- Gender:Unisex
- Closure type:Zip closure
- Water resistance:First line barrier
- Tensile strength:13 lbs./in. (warp), 16.1 lbs./in. (filling)
- Weight:50 g/m²
- Bacterial filtration efficiency:95.30%
- Tearing strength:3.1 lbs./in. (lengthwise Yarns), 2.6 lbs./in. (widthwise Yarns)
- Cat. No.:414004-331
- Cleanroom class:10000/ISO 7
- Air permeability:0.9 cu. ft./min./sq. ft.
- Garment application:Cleanroom|Industrial|Laboratory|General Purpose
- Pockets:Without pockets
- Fabric:Spunbound polypropylene
- Particle shedding (Helmke Drum):Level II
- Water vapor transmission rate:965 g/m²/24 hrs.



