Orders LinkContactUsLinkComponent
LPG Sample Cylinders, Koehler
1000 ml
Catalog # 37000-222
Supplier:   Koehler
 :  K27854
LPG Sample Cylinders, Koehler
Catalog # 37000-222
Supplier:  Koehler
LPG Sample Cylinders, Koehler
LPG Sample Cylinders, Koehler
1000 ml
Price
$10,453.12
Log in for availability
Each
Product Details & Documents
Welded stainless steel cylinders obtain representative samples of liquefied petroleum gases (LPG)
Two-valve type instruments feature a 20% outage tube and built-in pressure relief valve preset between 540 and 600psi (38 and 42kg/cm2).
Certifications: Conform to specifications: ASTM D1265, GPA 2140.
Specifications
  • Capacity:
    1000 ml