Order Entry
Canada
Orders LinkContactUsLinkComponent
 

Orion™ pH Buffer Bottles, Thermo Scientific

pH 4.01 buffer, 475 ml, Pink
  34182-306
 :  910104
Orion™ pH Buffer Bottles, Thermo Scientific
  34182-306
 :  
Orion™ pH Buffer Bottles, Thermo Scientific
Orion™ pH Buffer Bottles, Thermo Scientific

pH 4.01 buffer, 475 ml, Pink

 :  
 
$71.40
   
Each (475ml)
 
   
 

Perform accurate and reproducible pH calibrations with Orion™ pH buffer bottles.

  • All bottles feature pH/temperature tables printed on the labels for accurate calibrations and NIST traceability
  • Manufactured under ISO 9000 quality standards
  • Available in pint (475 ml) bottles or portable five packs of 2 oz. (5×60 ml) bottles

The pH 4.01, 5.00, 7.00, and 10.01 buffers are color-coded for ease of use. All Orion™ pH buffers are traceable to NIST standard reference materials.

 

 

  • Color:
    Pink
  • Description:
    pH 4.01 buffer
  • Size:
    475 ml

 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.