Order Entry
Canada
Orders LinkContactUsLinkComponent
TuffStuff™ Critical Task Wipers, Contec®

TuffStuff™ Critical Task Wipers, Contec®

TuffStuff™ heavy-duty wipes, pop-up dispenser box, 22.9×41.9 cm (9×16.5"), 100/box, 8 boxes/case
Catalog # 33502-722
Supplier:   CONTEC, INC. MX
Supplier #:  TSWC0917
TuffStuff™ Critical Task Wipers, Contec®
Catalog # 33502-722
Supplier:  CONTEC, INC. MX
TuffStuff™ Critical Task Wipers, Contec®
TuffStuff™ Critical Task Wipers, Contec®

TuffStuff™ heavy-duty wipes, pop-up dispenser box, 22.9×41.9 cm (9×16.5"), 100/box, 8 boxes/case

Price
$329.38
New Customer Promotional Price
Log in for availability
Case of 800
Product Details & Documents

TuffStuff™ Wipes are high-quality critical task wipes. Available in medium and heavy-duty formats, these wipes are solvent resistant and maintain their strength even when wet. TuffStuff™ wipes offer exceptional durability and cleanliness.

  • Hydroentangled nonwoven wipes are strong and durable without the use of binders
  • Medium and heavy-duty wipes available to match the ideal wipe for the specific application
  • Offers strength wet or dry, no residues or binders left behind
  • Highly sorbent without the use of surfactants
  • Multiple wipe sizes allow optimum selection for each job

TuffStuff™ Wipes are available in multiple sizes, weights, and packaging configurations to meet various customer needs. The expanded range includes TuffSat, consisting of heavy-duty dry wipes, quarter-folded in a tub that can be saturated with cleaning solution. This versatile option allows wipes to be customized with the correct cleaning solution for a particular environment.

Certifications: Recommended for ISO Class 6 to 8 environments.

Specifications
  • Description:
    TuffStuff™ heavy-duty wipes, pop-up dispenser box
  • Packaging:
    100/box, 8 boxes/case
  • W×L:
    22.9×41.9 cm (9×16.5")