Order Entry
Canada
Orders LinkContactUsLinkComponent
Blower Transition Adapters, Labconco®

Blower Transition Adapters, Labconco®

Blower Transition Adapter, 20.3 cm (8"), 1.3 kg (3 lbs.)
Catalog # 30182-130
Supplier:   Labconco
Supplier #:  4722400
Blower Transition Adapters, Labconco®
Catalog # 30182-130
Supplier:  Labconco
Blower Transition Adapters, Labconco®
Blower Transition Adapters, Labconco®

Blower Transition Adapter, 20.3 cm (8"), 1.3 kg (3 lbs.)

Price
$679.00
Log in for availability
Each
Product Details & Documents

Epoxy-coated steel blower transition adapters fit all Labconco coated steel blowers

Adapter allows you to connect thermoplastic duct to the exhaust side of a blower to create an exhaust stack. A nominal size PVC duct fits inside the opening.

Specifications
  • Description:
    Blower Transition Adapter
  • Shipping weight:
    1.3 kg (3 lbs.)
  • Nominal diameter:
    20.3 cm (8")