Elbows, Labconco
90° Elbows, 15.2 cm (6"), 34.3 cm (13½"), 12 (in ft. of straight duct), 3.6 kg (8 lbs.)
30152-640
: Labconco
: 4708700
PVC elbows, both 45° and 90°, are compatible with thermoplastic duct
Designed and engineered for quick installation and minimum pressure losses, elbows feature belled end connections to receive PVC duct directly.
- Ø:15.2 cm (6")
- Description:90° Elbows
- Height:34.3 cm (13½")
- Shipping weight:3.6 kg (8 lbs.)
- Equivalent resistance:12 (in ft. of straight duct)



