Corning® Pipette Tips, Microvolume
0.5 - 10 µl, -, Natural, 10 bags of 1000
Supplier: Corning
Supplier #: 4901
Jump to:
Product Details & Documents
These microvolume tips provide accurate, reliable performance and are designed for use with major brand ultra-micropipettors.
- All racked tips are certified RNase-/DNase-free and non pyrogenic
- Exceptional durability: all tip racks stand up to the demands of use with multi-channel pipettors and withstand repeated autoclaving cycles
Documents
Specifications
- Volume:0.5 - 10 µl
- Tip color:Natural
- Packed:10 bags of 1000
- Sterile:-