Corning® Pipette Tips, Microvolume
0.1 - 10 µl, +, Natural, Rack Packed
Supplier: Corning
Supplier #: 4894
Corning® Pipette Tips, Microvolume
Catalog # 29443-320
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
These microvolume tips provide accurate, reliable performance and are designed for use with major brand ultra-micropipettors.
- All racked tips are certified RNase-/DNase-free and non pyrogenic
- Exceptional durability: all tip racks stand up to the demands of use with multi-channel pipettors and withstand repeated autoclaving cycles
Documents
Specifications
- Volume:0.1 - 10 µl
- Tip color:Natural
- Packed:Rack Packed
- Sterile:+
- Cat. No.:29443-320
- Environmental certification:I-RECs, ISO 14001 - Reynosa
- Environmentally Preferable:Low Manufacturing Impact



