Orders LinkContactUsLinkComponent
Corning® Pipette Tips, Microvolume
0.5 - 10 µl, +, Natural, 10 Racks of 96
Catalog # 29443-300
Supplier:   Corning
 :  4830
Corning® Pipette Tips, Microvolume
Catalog # 29443-300
Supplier:  Corning
Corning® Pipette Tips, Microvolume
Corning® Pipette Tips, Microvolume
0.5 - 10 µl, +, Natural, 10 Racks of 96
Price
$108.00
New Customer Promotional Price
Log in for availability
Case of 960
Product Details & Documents
These microvolume tips provide accurate, reliable performance and are designed for use with major brand ultra-micropipettors.
  • All racked tips are certified RNase-/DNase-free and non pyrogenic
  • Exceptional durability: all tip racks stand up to the demands of use with multi-channel pipettors and withstand repeated autoclaving cycles
Specifications
  • Volume:
    0.5 - 10 µl
  • Tip color:
    Natural
  • Packed:
    10 Racks of 96
  • Sterile:
    +