Orders LinkContactUsLinkComponent
Corning® Pipette Tips, Microvolume
Corning® Pipette Tips, Microvolume
0.1 - 10 µl, -, Natural, Rack Packed
Catalog # 29443-298
Product available on GSA Advantage®
   Low Manufacturing Impact
Supplier:   Corning
Supplier #:  4826
Corning® Pipette Tips, Microvolume
Catalog # 29443-298
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
Corning® Pipette Tips, Microvolume
Corning® Pipette Tips, Microvolume
0.1 - 10 µl, -, Natural, Rack Packed
Price
$93.00
New Customer Promotional Price
Log in for availability
Case of 960
Product Details & Documents
These microvolume tips provide accurate, reliable performance and are designed for use with major brand ultra-micropipettors.
  • All racked tips are certified RNase-/DNase-free and non pyrogenic
  • Exceptional durability: all tip racks stand up to the demands of use with multi-channel pipettors and withstand repeated autoclaving cycles
Specifications
  • Volume:
    0.1 - 10 µl
  • Tip color:
    Natural
  • Packed:
    Rack Packed
  • Sterile:
    -
  • Cat. No.:
    29443-298
  • Environmental certification:
    I-RECs, ISO 14001 - Reynosa
  • Environmentally Preferable:
    Low Manufacturing Impact