Netwell™ Inserts, Sterile, Corning
6, 24 mm, 440 µm, 6/Plate
Supplier: Corning
Supplier #: 3480
Netwell™ Inserts, Sterile, Corning
Catalog # 29442-138
Supplier: Corning
Product available on GSA Advantage®
Jump to:
Product Details & Documents
Polyester mesh bottoms attached to PS rings or housing. Netwell™ inserts are used as tissue carriers, supports and strainers of small organs, tissue slices or explants at the air-media interface.
- Handy carrier for immunocytochemical staining of tissue slices
- Provides coarse filtration of tissue homogenates, cell suspensions and microcarriers
- Available in two mesh sizes and diameters
- Supplied sterile and pre-loaded in 6- or 12-well plates
- 24 mm Netwell inserts fit in Corning® 50 ml plastic centrifuge tubes
Polystyrene reagent trays are available in black for improved visibility of tissue sections, or white for colorimetric reaction contrast. Specially designed carriers and handles permit the simultaneous processing of up to 12 samples per carrier. Each carrier kit contains eight carriers and eight handles.
Specifications
- No. of wells:6
- Packaging:6/Plate
- Mesh size:440 µm
- Membrane diameter:24 mm
- Cat. No.:29442-138
- Environmental certification:RECs, ISO 14001 - Kennebunk
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Product Information
- Catalog #Membrane DiameterAvailabilityPrice
- Carrier kit, 6-well
Product Information
- Catalog #AvailabilityPrice
- Reagent Tray, Black
Product Information
- Catalog #Membrane DiameterAvailabilityPrice
- Carrier kit, 12-well
Product Information
- Catalog #AvailabilityPrice
- Reagent Tray, White
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



